Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H1N1 HA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520713
Description
This antibody detects a peptide sequence that differs from Product # PA5-20712 and PA5-20715, and can be used as a cognate pair with Product # PA5-20714. PA5-20713 can be used with blocking peptide PEP-0828.
Influenza A H1N1 HA Polyclonal antibody specifically detects Influenza A H1N1 HA in Virus samples. It is validated for ELISA.Specifications
Influenza A H1N1 HA | |
Polyclonal | |
Unconjugated | |
flu hemagglutinin; H1N1 HA; H1N1 hemagglutinin; Influenza H1N1; influenza hemagglutinin | |
A synthetic peptide from the Hemagglutinin protein. | |
100 μg | |
Primary | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
ELISA | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction