Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524231

Catalog No. PIPA524231

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene belongs to the SEC13 family of WD-repeat proteins. It is a constituent of the endoplasmic reticulum and the nuclear pore complex. It has similarity to the yeast SEC13 protein, which is required for vesicle biogenesis from endoplasmic reticulum during the transport of proteins. Multiple alternatively spliced transcript variants have been found.
Specifications

Specifications

SEC13
Polyclonal
PBS with 0.09% sodium azide
P55735
D3S1231E, SEC13L1, SEC13R, npp-20, SEC13-like 1 isoform, SEC13-like protein 1, SEC13-related protein, protein SEC13 homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6396
Flow Cytometry, Western Blot
Unconjugated
SEC13
Liquid
SEC13
KLH conjugated synthetic peptide between 72-100 amino acids from the central region of human SEC13
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit