Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC22B Sheep anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen OSS00071G

Catalog No. PIOSS00071G


Add to cart

Description

Description

Glycerol (1:1) may be added for additional stability. Reconstitute in 500 µL of sterile water.

The protein encoded by this gene is a member of the SEC22 family of vesicle trafficking proteins. It seems to complex with SNARE and it is thought to play a role in the ER-Golgi protein trafficking. This protein has strong similarity to Mus musculus and Cricetulus griseus proteins.
Specifications

Specifications

SEC22B
Polyclonal
Unconjugated
SEC22B
Lyophilized
SEC22B
A synthetic peptide from human SEC22b conjugated to an immunogenic carrier protein was used as the antigen
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry, Western Blot
1 mg/mL
Antibody with no preservative
O75396, O08547, Q4KM74
SEC22 vesicle-trafficking protein homolog B, SEC22 vesicle-trafficking protein-like 1, ERS24, ERS-24, Vesicle-trafficking protein SEC22b, SEC22L1, SEC22B
Sheep
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
20333, 310710, 9554
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit