Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sec31A Mouse anti-Human, Unlabeled, Clone: 32, BD

Mouse Monoclonal Antibody

$221.00 - $386.00

Specifications

Antigen Sec31A
Isotype IgG1
Immunogen Human Sec31A aa. 522-719
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB612350
View Documents Promotion Details
BD Biosciences
612350
50μg
Each for $221.00
Add to cart
 
BDB612351
View Documents Promotion Details
BD Biosciences
612351
150μg
Each for $386.00
Add to cart
 
Description

Description

Eukaryotic protein trafficking involves packaging of target molecules into membranous vesicles that bud from a donor compartment, travel to a specific destination, fuse, and release their contents into an acceptor compartment. Vesicles that bud from the Golgi cisternae and from the ER contain a non-clathrin based coat. This coat is an oligomeric complex whose subunits are termed COPs (coatomer proteins). COPI is the golgi- and endosome-associated COP complex, while COPII is the ER-associated coat complex. In yeast, the COPII complex has a cargo binding subcomplex that includes Sar1p, Sec23p, and Sec24p, which recruits Sec13p and Sec31p leading to vesicle formation. The human homologues of Sec31p are Sec31A and Sec31B. Sec31A has 40% identity with Sec31B, and contains an N-terminal WD-40 domain and a C-terminal proline-rich region. Sec31A mRNA is widely expressed, while Sec31B is found only in testis and thymus. In HeLa, Sec31A localizes to vesicular structures in the perinuclear region of the cell, and co-localizes with the COPII component Sec13. Antibodies against Sec31A inhibit ER to Golgi transport of vesicular stomatitis G protein. Thus, Sec31 may be an important COPII component involved in ER to Golgi transport, as well as a marker for COPII vesicles.

Immunofluorescence, Western Blotting

Specifications

Specifications

Sec31A
Human Sec31A aa. 522-719
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
32
250μg/mL
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit