Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sec61α Rabbit anti-Canine, Human, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07204
Description
Detect Sec61α using this Anti-Sec61α Antibody validated for use in immunoprecipitation and western blotting.Specifications
Sec61α | |
Polyclonal | |
antiserum containing 0.05% sodium azide and 30% glycerol | |
Rabbit | |
Antiserum | |
Primary | |
Canine, Human, Rat | |
Serum |
Immunoprecipitation, Western Blot | |
Unconjugated | |
SEC61A1; HSEC61; SEC61A; SEC61 | |
peptide conjugated to KLH and to BSA, corresponding to amino acids 463-476 of human Sec61a (C-KEQSEVGSMGALLF) | |
100 μL | |
NM_013336, NM_018144 | |
−20°C for 24 months | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction