Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sec8 Mouse, Unlabeled, Clone: 14, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610659

 View more versions of this product

Catalog No. BDB610659


Add to cart

Description

Description

Signal transmission between neurons is regulated by the release of neurotransmitters at the synapse. This process, which is controlled by a complex pathway of membrane trafficking in the presynaptic nerve terminal, leads to membrane fusion and neurotransmitter secretion. Sec8 is a hydrophilic protein of 975 amino acids that is highly expressed in brain and kidney. It is homologus with the yeastsecretory protein, Sec8p. In yeast, Sec8 is essential for the Golgi-to-plasma membrane traffic of proteins during constitutive secretion. In rat brain, Sec8 colocalizes with Rab3 and Syntaxin-1a, two important proteins that regulate the fusion of the synaptic vesicle to the plasma membrane. The primary structure of Sec8, a coiled-coil domain (residues 34-99), is commonly found in other proteins important in secretory pathways.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

Sec8
14
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.