Learn More
Secobarbital Sheep anti-Chemical, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA175403
Catalog No. PIPA175403
Add to cart
Description
Description
PA1-75403 specifically detects Secobarbital. PA1-75403 has been successfully used in ELISA applications. The PA1-75403 immunogen is Secobarbital(5)-BTG.
Secobarbital is a barbiturate derivative drug only found in individuals that have used the drug. It possesses anaesthetic, anticonvulsant, sedative and hypnotic properties. In the United Kingdom, it was known as Quinalbarbitone. Secobarbital binds at a distinct binding site associated with a Cl- ionopore at the GABAA receptor, increasing the duration of time for which the Cl- ionopore is open. The post-synaptic inhibitory effect of GABA in the thalamus is, therefore, prolonged. [National Center for Biotechnology Information. PubChem Compound Database; CID=5193]
Specifications
Specifications
Secobarbital
Polyclonal
PBS with 0.09% sodium azide
Quinalbarbitone; Seconal; Meballymal; Secobarbitone; Quinalbarbital; C12H18N2O3
Secobarbital(5)-BTG
Purified
RUO
Antibody
Polyclonal
ELISA
Unconjugated
Liquid
Sheep
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical