Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLPI Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA135993

Catalog No. 01-674-551


Add to cart

Description

Description

This gene encodes a secreted inhibitor which protects epithelial tissues from serine proteases. It is found in various secretions including seminal plasma, cervical mucus, and bronchial secretions, and has affinity for trypsin, leukocyte elastase, and cathepsin G. Its inhibitory effect contributes to the immune response by protecting epithelial surfaces from attack by endogenous proteolytic enzymes; the protein is also thought to have broad-spectrum anti-biotic activity.
Specifications

Specifications

SLPI
Polyclonal
Unconjugated
SLPI
Liquid
SLPI
Human SLPI.
Purified
RUO
Antibody
Polyclonal
Human
ELISA, Immunoprecipitation, Western Blot
0.1 mg/mL
PBS with 0.1% BSA and 0.02% sodium azide
P03973
ALK1, ALP, BLPI, HUSI, HUSI-I, MPI, WAP4, WFDC4
Rabbit
IgG
100 μg
4° C
Primary
6590
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit