Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA6A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528187

Catalog No. PIPA528187


Add to cart

Description

Description

PA5-28187 targets SEMA6A in IHC (P), ICC and WB applications and shows reactivity with Human and mouse samples. The PA5-28187 immunogen is recombit fragment corresponding to a region within amino acids 247 and 534 of Human SEMA6A. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Can act as repulsive axon guidance cues. May play a role in channeling sympathetic axons into the sympathetic chains and controlling the temporal sequence of sympathetic target innervation.
Specifications

Specifications

SEMA6A
Polyclonal
Unconjugated
SEMA6A
Liquid
SEMA6A
Recombit fragment corresponding to a region within amino acids 247 and 534 of Human SEMA6A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9H2E6, O35464
HT018, SEMA, SEMA6A1, SEMAQ, VIA, SEMA6A-1, sema VIa, semaphorin 6A-1, semaphorin VIA, semaphorin-6A, semaphorin-6A-1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
20358, 57556
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit