Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Semaphorin 4A, PE, Clone: 757129, R&D Systems™

Manufacturer:  R&D Systems FAB8166P

Catalog No. FAB8166P

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

This item is not returnable. View return policy

Description

Description

Host Species: Rat
Clone: 757129
Isotype: IgG1
Species Reactivity: Ms
Immunogen: Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 4A, Thr33-Trp638

Applications:

Flow Cytometry

Specifications

Specifications

Semaphorin 4A
757129
Flow Cytometry 10 uL/10^6 cells
Q62178
CORD10, RP35, Sema B, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4A, SEMA4A, SEMAB, semaphorin-4A, Semaphorin-B, SemB, SEMBFLJ12287
Rat
IgG1
100 Tests
Primary
64218
Mouse
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
SEMA4A
Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 4A, Thr33-Trp638, Accession # Q62178
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects mouse Semaphorin 4A in ELISA. Stains HEK293 cells transfected with mouse Semaphorin 4A by Flow Cytometry, but does not stain untransfected cells.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit