Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA5A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526066

Catalog No. PIPA526066

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with rat based on sequence homology.

Semaphorin 5A belongs to the semaphorin gene family that encodes membrane proteins containing a semaphorin domain and several thrombospondin type-1 repeats. Members of this family are involved in axonal guidance during neural development.
Specifications

Specifications

SEMA5A
Polyclonal
PBS with 0.09% sodium azide
Q13591, Q62217
SEMAF, semF, sema F, semaphorin F, semaphorin-5A, semaphorin-F
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
20356, 9037
Western Blot
Unconjugated
SEMA5A
Liquid
SEMA5A
KLH conjugated synthetic peptide between 90-113 amino acids from human Semaphorin 5A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit