Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA6D Mouse anti-Human, Clone: 257510, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523955

Catalog No. PIMA523955


Add to cart

Description

Description

In Western blots, approximately 10% cross-reactivity with recombit human (rh) Semaphorin 3A and recombit mouse (rm) Semaphorin 3C is observed and no cross-reactivity with rhSemaphorin 3B, 6A, 6B, 6C, 7A, rmSemaphorin 3E, 3F, or 6D is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

SEMA6D
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q8NFY4
Sema6D, KIAA1479FLJ11598, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6D, semaphorin-6D
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
80031
Western Blot
257510
Unconjugated
SEMA6D
Lyophilized
SEMA6D
Mouse myeloma cell line NS0-derived recombit human Semaphorin 6D Ser22-His660
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit