Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUMO1/sentrin specific peptidase 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human SENP1 protein.

Manufacturer:  Abnova Corporation H00029843B01

Catalog No. 89-007-504


Add to cart

Description

Description

The covalent modification of proteins by the small ubiquitin (UBB; MIM 191339)-like protein SUMO (see SUMO1, MIM 601912) is implicated in the regulation of nucleocytoplasmic transport, genomic stability, gene transcription, and other processes. Sumoylation is catalyzed on target lysine residues by a multienzyme process and is reversed by desumoylating enzymes such as SENP1 (Yamaguchi et al., 2005 [PubMed 15923632]).[supplied by OMIM

Sequence: MDDIADRMRMDAGEVTLVNHNSVFKTHLLPQTGFPEDQLSLSDQQILSSRQGHLDRSFTCSTRSAAYNPSYYSDNPSSDSFLGSGDLRTFGQSANGQWRNSTPSSSSSLQKSRNSRSLYLETRKTSSGLSNSFAGKSNHHCHVSAYEKSFPIKPVPSPSWSGSCRRSLLSPKKTQRRHVSTAEETVQEEEREVYRQLLQMVTGKQFTIAKPTTHFPLHLSRCLSSSKNTLKDSLFKNGNSCASQIIGSDTSSSGSASILTNQEQLSHSVYSLSSYTPDVAFGSKDSGTLHHPHHHHSVPHQPDNLAASNTQSEGSDSVILLKVKDSQTPTPSSTFFQAELWIKELTSVYGSRARERLRQIEEQKALALQLQNQRLQEREHSVHDSVELHLRVPLEKEIPVTVVQETQKKGHKLTDSEDEFPEITEEMEKEIKNVFRNGNQDEVLSEAFRLTITRKDIQTLNHLNWLNDEIINFYMNMLMERSKEKGLPSVHAFNTFFFTKLKTAGYQAVKRWTKKVDVFSVDILLVPIHLGVHWCLAVVDFRKKNITYYDSMGGINNEACRILLQYLKQESIDKKRKEFDTNGWQLFSKKSQEIPQQMNGSDCGMFACKYADCITKDRPINFTQQHMPYFRKRMVWEILHRKLL
Specifications

Specifications

SUMO1/sentrin specific peptidase 1
Unconjugated
No additive
BC045639.1
SuPr-2
Mouse
50 uL
Ubiquitin/Proteasome
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human SENP1 protein.
SENP1
Serum
SENP1
SENP1 (AAH45639.2, 1 a.a. ~ 644 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
29843
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit