Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SENP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511327

Catalog No. PIPA511327


Add to cart

Description

Description

SENP1 is a protease that catalyzes two essential functions in the SUMO pathway: processing of full-length SUMO1, SUMO2 and SUMO3 to their mature forms and deconjugation of SUMO1, SUMO2 and SUMO3 from targeted proteins. SENP deconjugates SUMO1 from HIPK2 and from HDAC1, which decreases the transcriptional repression activity of the latter.
Specifications

Specifications

SENP1
Polyclonal
PBS with 0.09% sodium azide
Q9P0U3
Sentrin-specific protease 1; Sentrin/SUMO-specific protease SENP1; SEN1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29843
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SENP1
Liquid
SENP1
KLH conjugated synthetic peptide between 28-59 amino acids from the N-terminal region of human SENP1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit