Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SENP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP131217
Description
SENP2 Polyclonal specifically detects SENP2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SENP2 | |
Polyclonal | |
Unconjugated | |
PBS, 1% BSA, 20% Glycerol (pH7) with 0.01% Thimerosal | |
SENP2 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human SENP2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
59343 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
AXAM2Axam2, DKFZp762A2316, EC 3.4.22, EC 3.4.22.68, KIAA1331sentrin-specific protease 2, Sentrin/SUMO-specific protease SENP2, Smt3ip2, SMT3IP2EC 3.4.22.-, SMT3-specific isopeptidase 2, SUMO1/sentrin/SMT3 specific peptidase 2, SUMO1/sentrin/SMT3 specific protease 2 | |
Rabbit | |
68 kDa | |
100 μL | |
Primary | |
Tested in human. Predicted cross-reactivity based on sequence identity: Bovine (88%). Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction