Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SENP5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511335

Catalog No. PIPA511335

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and non-human primate based on sequence homology.

SENP5 is a protease that catalyzes two essential functions in the SUMO pathway: processing of full-length SUMO3 to its conjugatable mature form and deconjugation of SUMO2 and SUMO3 from targeted substrates. This protein has weak proteolytic activity against full-length SUMO1 or SUMO1 conjugates. SENP5 is required for cell division.
Specifications

Specifications

SENP5
Polyclonal
PBS with 0.09% sodium azide
Q96HI0
Sentrin-specific protease 5; Sentrin/SUMO-specific protease SENP5; FKSG45
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
205564
Immunohistochemistry, Western Blot
Unconjugated
SENP5
Liquid
SENP5
KLH conjugated synthetic peptide between 524-555 amino acids from the C-terminal region of human SENP5
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit