Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Septin-6 Mouse anti-Human, Canine, Clone: 9E7, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051566
Description
Anti-Septin-6 Antibody, clone 9E7 is a Mouse Monoclonal Antibody for the detection of Septin-6 and has been validated in western blotting and immunoprecipitation.Specifications
Septin-6 | |
Monoclonal | |
Unconjugated | |
NP_055944 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG3 κ |
Immunoprecipitation, Western Blot | |
9E7 | |
Purified mouse monoclonal IgG3κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
SEPT6, KIAA0128, SEP2, SEPT2 | |
GST-tagged recombinant protein corresponding to human Septin-6. | |
100 μL | |
Canine, Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only