Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERCA3 ATPase, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1910A

Catalog No. PA1910A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ATP dependent calcium pumps are responsible in part for the maintenance of low cytoplasmic free calcium concentrations. The ATP pumps that reside in intracellular organelles are encoded by a family of structurally related enzymes, termed the sarcoplasmic or endoplasmic reticulum calcium ATPases (SERCA). The SERCA2 gene is subject to tissue dependent processing which is responsible for the generation of SERCA2a muscle-specific isoform expressed in type I (slow) skeletal, cardiac and smooth muscle and the SERCA2b isoform expressed in all cell types. SERCA3 has been found to co-express with SERCA2b in platelets, mast cells, lymphoid cells, and epithelial cells. Due to this pattern of expression it is referred to as the non-muscle form of SERCA.

PA1-910A detects sarcoplasmic or endoplasmic reticulum calcium ATPase 3 (SERCA3) mouse and rat samples. This antibody does not detect other SERCA isoforms.PA1-910A has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~97kDa protein representing recombinant SERCA3 from rat samples.PA1-910A immunizing peptide corresponds to amino acid residues 29-39 from rat and mouse SERCA3. PA1-910A immunizing peptide (Cat. # PEP-015) is available for neutralization and control experiments.
Specifications

Specifications

SERCA3 ATPase
Polyclonal
Unconjugated
Atp2a3
Liquid
ATP2A3
Synthetic Peptide: V(29) T D A R E R Y G P N(39).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Murine, Rat
Western Blot
1mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q64518, P18596
Sarcoplasmic or Endoplasmic Reticulum Calcium ATPase 3
Rabbit
IgG
100μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25391, 53313
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit