Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serine protease 23 Rabbit anti-Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP179488

Catalog No. NBP179488


Add to cart

Description

Description

Serine protease 23 Polyclonal antibody specifically detects Serine protease 23 in Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish samples. It is validated for Western Blot.
Specifications

Specifications

Serine protease 23
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
PRSS23
Synthetic peptide directed towards the C terminal of human PRSS23. Peptide sequence VSPPAKQLPGGRIHFSGYDNDRPGNLVYRFCDVKDETYDLLYQQCDAQPG.
41 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Western Blot
Western Blot 1:1000
NP_009104
EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, MGC5107, protease, serine, 23, Putative secreted protein Zsig13, serine protease 23, serine protease, umbilical endothelium, SIG13, SPUVE, ZSIG13
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
11098
Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only