Learn More
Serotonin Rabbit anti-Chemical, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136157
Catalog No. 01-672-110
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 μL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide. For long-term storage, aliquot and store at -20°C.
Serotonin (also known as 5-hydroxytryptamne or 5-HT) is a biochemical messenger and neurotransmitter synthesized from L-tryptophan. Serotonin has a range of functions, including regulation of gastric motility, and may be related to some psychiatric disorders including schizophrenia and depression. Serotoinin is widely distributed in the central and peripheral nervous system, and also in some non-nueral tissues such as platelets.
Specifications
Specifications
Serotonin
Polyclonal
whole serum
5-HT; 5-hydroxytryptamne
Serotonin conjugated to BSA.
50 μL
4° C
Primary
Chemical, Rat
Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
Lyophilized
Rabbit
IgG
RUO
Antibody
Polyclonal