Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTR1D Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141164

Catalog No. 01-675-738


Add to cart

Description

Description

Suggested positive control: human, mouse, or rat brain.

The 5-HT1D receptor is a member of the Serotonin Receptor subfamily. 5-HT1D receptor expression has been documented primarily in brain and in dorsal root ganglion, heart, and small intestine. ESTs have been isolated from human brain and eye libraries.
Specifications

Specifications

HTR1D
Polyclonal
Unconjugated
HTR1D
Liquid
HTR1D
Synthetic peptides corresponding to residues 1-18 and 251-267 of rat 5-HT1DR.
Protein G
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P28221, Q61224, P28565
5-HT1DR
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
15552, 25323, 3352
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit