Learn More
anti-Serpin A10/ZPI, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP130808
Catalog No. NBP130808
Add to cart
Description
Description
Serpin A10/ZPI Polyclonal antibody specifically detects Serpin A10/ZPI in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Serpin A10/ZPI
LYOPH
Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Antisera
SERPINA10
A synthetic peptide from n-terminal region of human Serpin A10 (SERPINA10) conjugated to an immunogenic carrier protein was used as the immunogen.
Unpurified
RUO
Primary
51156
Reconstitute with 0.1 ml sterilized water to desired concentration.
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Whole antisera with No Preservative
antitrypsin), member 10, member 10, PZIprotein Z-dependent protease inhibitor, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A10, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), ZPIPZ-dependent protease inhibitor
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Appears to be specific for Serpin A10.
Human