Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Serpin A10/ZPI, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132903

Catalog No. NBP132903


Add to cart

Description

Description

Serpin A10/ZPI Polyclonal antibody specifically detects Serpin A10/ZPI in Human samples. It is validated for Western Blot.
Specifications

Specifications

Serpin A10/ZPI
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
SERPINA10
Recombinant protein encompassing a sequence within the center region of human SERPINA10. The exact sequence is proprietary.
51 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
antitrypsin), member 10, member 10, PZIprotein Z-dependent protease inhibitor, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A10, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), ZPIPZ-dependent protease inhibitor
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
51156
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only