Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINA12 Mouse anti-Human, Clone: 484223, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524197

Catalog No. PIMA524197


Add to cart

Description

Description

In Western blots, no cross-reactivity with recombit human (rh) Serpin A1, A3, A4, A5, A6, B8, 9, C1, D1, E1, F2, G1, I1, I2, recombit mouse (rm) Serpin E2, F1, recombit rat Serpin B6, rhA1 Antitrypsin, rhAngiotensinogin, or rmPEDF is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

May modulates insulin action conceivably only in the presence of its yet undefined target proteases in white adipose tissues.
Specifications

Specifications

SERPINA12
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q8IW75
OL-64, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 12, serpin A12, vaspin, visceral adipose tissue-derived serine protease inhibitor, visceral adipose-specific SERPIN
Mouse
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
145264
Immunoprecipitation, Western Blot
484223
Unconjugated
SERPINA12
Lyophilized
SERPINA12
Mouse myeloma cell line NS0-derived recombit human Serpin A12 Leu20-Lys414
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit