Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Serpin B8/Proteinase Inhibitor 8, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP145799

Catalog No. NBP145799

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Serpin B8/Proteinase Inhibitor 8
Unconjugated
PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.02% Thimerosal
Affinity Purified
SERPINB8
Synthetic peptide surrounding amino acid 10 of human PI-8.
Immunogen affinity purified
RUO
Primary
5271
Western Blot
Western Blot
P50452
CAP-2, CAP2clade B (ovalbumin), member 8, Cytoplasmic antiproteinase 2, Peptidase inhibitor 8, PI-8, protease inhibitor 8 (ovalbumin type), serpin B8, serpin peptidase inhibitor, clade B (ovalbumin), member 8
Rabbit
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only