Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-1 Antitrypsin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526439

Catalog No. PIPA526439


Add to cart

Description

Description

This antibody is predicted to react with mouse, rat and Xenopus based on sequence homology.

SERPINA1 is secreted and is a serine protease inhibitor whose targets include elastase, plasmin, thrombin, trypsin, chymotrypsin, and plasminogen activator. Defects in this gene can cause emphysema or liver disease.
Specifications

Specifications

alpha-1 Antitrypsin
Polyclonal
PBS with 0.09% sodium azide
P01009
A1A, A1AT, AAT, PI, PI1, PRO2275, alpha1AT, alpha-1 protease inhibitor, alpha-1-antiproteinase, alpha-1-antitrypsin, alpha-1-antitrypsin null, protease inhibitor 1 (anti-elastase), alpha-1-antitrypsin, serine (or cysteine) proteinase inhibitor, clade A, member 1, serpin A1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5265
Western Blot
Unconjugated
SERPINA1
Liquid
SERPINA1
KLH conjugated synthetic peptide between 158-187 amino acids from the central region of human SERPINA1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit