Learn More
SERPINA3, Rabbit, MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human SERPINA3 protein.
Supplier: Abnova Corporation H00000012D01
Description
The protein encoded by this gene is a plasma protease inhibitor and member of the serine protease inhibitor class. Polymorphisms in this protein appear to be tissue specific and influence protease targeting. Variations in this protein's sequence have been implicated in Alzheimer's disease, and deficiency of this protein has been associated with liver disease. Mutations have been identified in patients with Parkinson disease and chronic obstructive pulmonary disease. [provided by RefSeq]
Sequence: MERMLPLLALGLLAAGFCPAVLCHPNSPLDEENLTQENQDRGTHVDLGLASANVDFAFSLYKQLVLKAPDKNVIFSPLSISTALAFLSLGAHNTTLTEILKGLKFNLTETSEAEIHQSFQHLLRTLNQSSDELQLSMGNAMFVKEQLSLLDRFTEDAKRLYGSEAFATDFQDSAAAKKLINDYVKNGTRGKITDLIKDLDSQTMMVLVNYIFFKAKWEMPFDPQDTHQSRFYLSKKKWVMVPMMSLHHLTIPYFRDEELSCTVVELKYTGNASALFILPDQDKMEEVEAMLLPETLKRWRDSLEFREIGELYLPKFSISRDYNLNDILLQLGIEEAFTSKADLSGITGARNLAVSQVVHKAVLDVFEEGTEASAATAVKITLLSALVETRTIVRFNRPFLMIIVPTDTQNIFFMSKVTNPKQASpecifications
SERPINA3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human SERPINA3 protein. | |
SERPINA3 | |
AACT/ACT/GIG24/GIG25/MGC88254 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
No additive | |
NM_001085.4 | |
SERPINA3 | |
SERPINA3 (NP_001076.2, 1 a.a. ∼ 423 a.a) full-length human protein. | |
RUO | |
12 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.