Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINA4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514359

Catalog No. PIPA514359


Add to cart

Description

Description

SERPINA4 inhibits human amidolytic and kininogenase activities of tissue kallikrein. The inhibition is achieved by formation of an equimolar, heat- and SDS-stable complex between the inhibitor and the enzyme, and generation of a small C-terminal fragment of the inhibitor due to cleavage at the reactive site by tissue kallikrein.
Specifications

Specifications

SERPINA4
Polyclonal
PBS with 0.09% sodium azide
P29622
Kallistatin; Kallikrein inhibitor; Protease inhibitor 4; Serpin A4 ; PI4; synonyms: KST; name: SERPINA4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5267
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
SERPINA4
Liquid
SERPINA4
KLH conjugated synthetic peptide between 291-320 amino acids from the C-terminal region of human SERPINA4
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit