Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
serpin peptidase inhibitor, clade B (ovalbumin), member 3 (D03P), Rabbit anti-Human, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human SERPINB3 protein.
Supplier: Abnova Corporation H00006317D03P
Description
Sequence: MNSLSEANTKFMFDLFQQFRKSKENNIFYSPISITSALGMVLLGAKDNTAQQIKKVLHFDQVTENTTGKAATYHVDRSGNVHHQFQKLLTEFNKSTDAYELKIANKLFGEKTYLFLQEYLDAIKKFYQTSVESVDFANAPEESRKKINSWVESQTNEKIKNLIPEGNIGSNTTLVLVNAIYFKGQWEKKFNKEDTKEEKFWPNKNTYKSIQMMRQYTSFHFASLEDVQAKVLEIPYKGKDLSMIVLLPNEIDGLQKLEEKLTAEKLMEWTSLQNMRETRVDLHLPRFKVEESYDLKDTLRTMGMVDIFNGDADLSGMTGSRGLVLSGVLHKAFVEVTEEGAEAAAATAVVGFGSSPTSTNEEFHCNHPFLFFIRQNKTNSILFYGRFSSPSpecifications
serpin peptidase inhibitor, clade B (ovalbumin), member 3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human SERPINB3 protein. | |
SERPINB3 | |
HsT1196/SCC/SCCA-1/SCCA-PD/SCCA1/T4-A | |
Rabbit | |
Affinity Purified | |
RUO | |
6317 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC005224.1 | |
SERPINB3 | |
SERPINB3 (AAH05224, 1 a.a. ∼ 390 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction