Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAI1 Mouse anti-Mouse, Rat, Clone: MA-124K1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA140249

Catalog No. 01-676-582

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PAI-1 mediates inhibition of fibrinolysis by inhibiting the activity of plasminogen activator; may promote neuronal survival.

MA1-40249 detects PAI-1 from rat and mouse samples.MA1-40249 has been successfully used in ELISA and functional assay applications.The MA1-40249 immunogen is Glu220/Glu212 of PAI-1.
Specifications

Specifications

PAI1
Monoclonal
0.1 mg/mL
PBS with 0.1% BSA and no preservative
P22777, P20961
SERPINE1
Mouse
IgG1
100 μg
4° C
Primary
18787, 24617
ELISA, Functional Assay
MA-124K1
Unconjugated
Serpine1
Liquid
SERPINE1
Glu220/Glu212 of PAI-1.
Purified
RUO
Antibody
Monoclonal
Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit