Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serum Albumin Chicken anti-Human, Biotin, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA172057

Catalog No. PA172057


Add to cart

Description

Description

PA1-72057 detects human serum albumin in human samples. PA1-72057 has been successfully used in ELISA and western blot procedures. The PA1-72057 immunogen is from purified normal human serum albumin.

Albumin is a soluble, monomeric protein with a molecular weight of approximately 65-67 kDa that comprises about one-half of the blood serum protein. Human serum albumin (HAS) is the most abundant protein in human blood plasma, and has a serum half-life of approximately 20 days. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular, unglycosylated serum protein synthesized in the liver as pre-pro-albumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough `produce the secreted albumin. Serum albumin has a good binding capacity for water, Ca(2+), Na(+), K(+), fatty acids, hormones, bilirubin and drugs. The main function of albumin is the regulation of the colloidal osmotic pressure of blood.
Specifications

Specifications

Human Serum Albumin
Polyclonal
Biotin
ALB
Liquid
ALB
Purified normal human serum albumin.
Purified
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
1 mg/mL
PBS with 50% glycerol and 0.01% thimerosal
P02768
Albumin, HSA, Serum albumin precursor, ALB
Chicken
IgY
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
213
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit