Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
sestrin 3, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human SESN3 protein.
Supplier: Abnova Corporation H00143686B01P
Description
Sequence: MNRGGGSPSAAANYLLCTNCRKVLRKDKRIRVSQPLTRGPSAFIPEKEVVQANTVDERTNFLVEEYSTSGRLDNITQVMSLHTQYLESFLRSQFYMLRMDGPLPLPYRHYIAIMAAARHQCSYLINMHVDEFLKTGGIAEWLNGLEYVPQRLKNLNEINKLLAHRPWLITKEHIQKLVKTGENNWSLPELVHAVVLLAHYHALASFVFGSGINPERDPEISNGFRLISVNNFCVCDLANDNNIENASLSGSNFGIVDSLSELEALMERMKRLQEEREDEEASQEEMSTRFEKEKKESLFVVSGDTFHSFPHSGAFLHFFAFSpecifications
sestrin 3 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human SESN3 protein. | |
SESN3 | |
MGC29667/SEST3 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Immunofluorescence, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC017296 | |
SESN3 | |
SESN3 (AAH17296.1, 1 a.a. ∼ 321 a.a) full-length human protein. | |
50 μg | |
Cell Cycle | |
143686 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction