Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SET Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521756

Catalog No. PIPA521756


Add to cart

Description

Description

The PA5-21756 immunogen is a synthetic peptide corresponding to a region within amino acids 1 and 45 of SET. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

SET
Polyclonal
Unconjugated
SET
Liquid
SET
Synthetic peptide corresponding to a region within amino acids 1 and 45 of SET
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.959 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q01105
RP11-216B9.3, 2PP2A, I2PP2A, IGAAD, IPP2A2, PHAPII, TAF-I, TAF-IBETA, HLA-DR-associated protein II, SET translocation (myeloid leukemia-associated), Template-Activating Factor-I, chromatin remodelling factor, inhibitor of granzyme A-activated DNase, inhibitor-2 of protein phosphatase-2A, phosphatase 2A inhibitor I2PP2A, protein SET, protein phosphatase type 2A inhibitor
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6418
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit