Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SET1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128685
Description
SET1B Polyclonal specifically detects SET1B in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).Specifications
SET1B | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
EC 2.1.1, FLJ20803, histone-lysine N-methyltransferase SETD1B, hSET1B, KIAA1076EC 2.1.1.43, KMT2GSET1B, Lysine N-methyltransferase 2G, SET domain containing 1B, SET domain-containing protein 1B, Set1B | |
Rabbit | |
Affinity Purified | |
RUO | |
23067 | |
Store at 4C. Do not freeze. |
Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
Immunohistochemistry 1:10-1:500, Immunoprecipitation 10 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500, Western Blot (Negative) | |
NP_055863.1 | |
SETD1B | |
The immunogen recognized by this antibody maps to a region between residue 1400 and 1450 of human SET domain containing 1B using the numbering given in entry NP_055863.1 (GeneID 23067). | |
100 μL | |
Primary | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction