Learn More
SETA/CIN85/Ruk/SH3KBPI Mouse anti-Human, Mouse, Rat, Clone: 179.1.E1, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05731
Catalog No. 05-731-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This Anti-SETA/CIN85/Ruk/SH3KBPI Antibody, clone 179.1.E1 is validated for use in immunoprecipitation, western blotting for the detection of SETA/CIN85/Ruk/SH3KBPI.
Specifications
Specifications
SETA/CIN85/Ruk/SH3KBPI
179.1.E1
of 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
κ
200μg
Primary
Human, Murine, Rat
Immunoprecipitation, Western Blot
Unconjugated
NM_031892
SH3KBP1, CD2BP3, OTTHUMP00000061480, HSB-1, GIG10, CIN85, MIG18
6XHIS and HA tagged fusion protein corresponding to full length rat SETA
Protein G purfied
−20°C for 24 months.
Monoclonal