Learn More
SETA/CIN85/Ruk/SH3KBPI Mouse anti-Human, Mouse, Rat, Clone: 84, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05733
Catalog No. 05-733-MI
Add to cart
Description
Description
Detect SETA/CIN85/Ruk/SH3KBPI using this Anti-SETA/CIN85/Ruk/SH3KBPI Antibody, clone 84 validated for use in immunoprecipitation, western blotting and immunocytochemistry.
Specifications
Specifications
SETA/CIN85/Ruk/SH3KBPI
84
Prtotein G purified antibody in 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4
Purified
Murine
IgG
200μg
Primary
Human, Murine, Rat
Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
NM_031892
SH3KBP1, CD2BP3, OTTHUMP00000061480, HSB-1, GIG10, CIN85, MIG18
6His and HA tagged fusion protein corresponding to full length rat SETA
Protein G chromatography
2-8°C for one year
Monoclonal