Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SETBP1 Rabbit anti-Human, Mouse, Porcine, Bovine, Equine, Rabbit, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP180331

Catalog No. NBP180331


Add to cart

Description

Description

SETBP1 Polyclonal antibody specifically detects SETBP1 in Human, Mouse, Porcine, Bovine, Equine, Rabbit samples. It is validated for Western Blot.
Specifications

Specifications

SETBP1
Unconjugated
PBS & 2% Sucrose. with 0.09% Sodium Azide
Affinity Purified
SETBP1
Synthetic peptide directed towards the middle region of human SETBP1. Peptide sequence TPRGGKRKHKPQAPAQPPQQSPPQQPLPQEEEVKAKRQRKSRGSESEVLP.
170 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Western Blot
Western Blot 1:1000
NP_056374
KIAA0437SEBDKFZp666J1210, SET binding protein 1, SET-binding protein
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
26040
Bovine, Equine, Human, Mouse, Porcine, Rabbit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only