Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SETD8, Biotin, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191269B

Catalog No. NBP191269B


Add to cart

Description

Description

SETD8 Polyclonal antibody specifically detects SETD8 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry (Negative), ICC/IF (Negative).
Specifications

Specifications

SETD8
Biotin
PBS with 0.05% Sodium Azide
Affinity Purified
SETD8
A synthetic peptide made to an internal portion of the human SETD8 protein (between residues 100-200) [UniProt Q9NQR1]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Mouse
Immunocytochemistry, Immunohistochemistry, Western Blot
Western Blot 1:1000, Immunohistochemistry (Negative), ICC/IF (Negative)
Q9NQR1
H4-K20-HMTase SETD8, H4K20-specific histone methyltransferase splice variant Set8b, Histone-lysine N-methyltransferase SETD8, KMT5A, KMT5A SET domain-containing protein 8, Lysine N-methyltransferase 5A, PR/SET domain containing protein 8, PR/SET domain-containing protein 07, PR/SET07, PRSET7, PR-Set7EC 2.1.1.43, SET domain containing (lysine methyltransferase) 8, SET07SET8H4-K20-specific histone methyltransferase, SET8
Rabbit
IgG
0.1 ml
Cell Cycle and Replication, Epigenetics
Primary
387893
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only