Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETDB2 Mouse anti-Mouse, Human, Clone: 5B7.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051952
Description
Use Anti-SETDB2 Antibody, clone 5B7.2 (mouse monoclonal antibody) validated in western blotting to detect SETDB2 also known as SET domain bifurcated 2, chronic lymphocytic leukemia deletion region 8, histone-lysine N-methyltransferase SETDB2.Specifications
SETDB2 | |
Monoclonal | |
Unconjugated | |
NP_001153780 | |
Mouse | |
Ion Exchange | |
Primary | |
2-8°C for one year | |
IgM κ |
Western Blot | |
5B7.2 | |
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide. | |
SETDB2, CLLD8, KMT1F, CLLL8 | |
Recombinant protein corresponding to human SETDB2. | |
100 μg | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only