Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEZ6L2 Rat anti-Mouse, Clone: 529805, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524231

Catalog No. PIMA524231


Add to cart

Description

Description

In direct ELISAs, no cross-reactivity with recombit mouse (rm) BSRP-B or rmBSRP-C is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

SE6L2 may contribute to specialized endoplasmic reticulum functions in neurons.
Specifications

Specifications

SEZ6L2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q4V9Z5
PSK-1, seizure 6-like protein 2, type I transmembrane receptor (seizure-related protein)
Rat
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
233878
Immunohistochemistry (Frozen), Western Blot
529805
Unconjugated
Sez6l2
Lyophilized
SEZ6L2
Mouse myeloma cell line NS0-derived recombit mouse SEZ6L2/BSRP-A Leu28-Ser837
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit