Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFMBT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522393

Catalog No. PIPA522393


Add to cart

Description

Description

PA5-22393 targets SFMBT1 in WB applications and shows reactivity with Human samples. The PA5-22393 immunogen is a recombit fragment corresponding to a region within amino acids 13 and 225 of SFMBT1 (Uniprot IDNo.Q9UHJ3). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

SFMBT1
Polyclonal
Unconjugated
SFMBT1
Liquid
SFMBT1
Recombit fragment corresponding to a region within amino acids 13 and 225 of SFMBT1 (Uniprot IDNo.Q9UHJ3)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.6 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q9UHJ3
RU1, SFMBT, Scm-related gene containing four mbt domains, Scm-related gene product containing four mbt domains, renal ubiquitous protein 1, scm-like with four MBT domains protein 1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
51460
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit