Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLASRP Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525270
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
SFRS16 is probably functions as an alternative splicing regulator. SFRS16 may regulate the mRNA splicing of genes such as CLK1. This may act by regulating members of the CLK kinase family.Specifications
CLASRP | |
Polyclonal | |
CLASRP | |
CLASP, SFRS16, SWAP2, Clk4 associating SR-related protein, splicing factor, arginine/serine-rich 16 (suppressor-of-white-apricot homolog, Drosophila), suppressor of white apricot homolog 2, suppressor of white-apricot homolog 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11129 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
Q8N2M8 | |
CLASRP | |
KLH conjugated synthetic peptide between 254-283 amino acids from the central region of human SFRS16 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction