Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGF29 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525064

Catalog No. PIPA525064

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with chicken, mouse and rat based on sequence homology.

CCDC101 is a subunit of 2 histone acetyltransferase complexes: the ADA2A (TADA2A; MIM 602276)-containing (ATAC) complex and the SPT3 (SUPT3H; MIM 602947)-TAF9 (MIM 600822)-GCN5 (KAT2A; MIM 602301)/PCAF (KAT2B; MIM 602303) acetylase (STAGA) complex. Both of these complexes contain either GCN5 or PCAF, which are paralogous acetyltransferases.
Specifications

Specifications

SGF29
Polyclonal
PBS with 0.09% sodium azide
Q96ES7
SGF29, STAF36, SAGA-associated factor 29 homolog, coiled-coil domain-containing protein 101
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
112869
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
SGF29
Liquid
SGF29
KLH conjugated synthetic peptide between 179-208 amino acids from the C-terminal region of human SGF29
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit