Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGIP1 Rabbit anti-Hamster, Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526562

Catalog No. PIPA526562

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with rat based on sequence homology.

SGIP1 functions as an endocytic protein that affects signaling by receptors in neuronal systems involved in energy homeostasis via its interaction with endophilins.
Specifications

Specifications

SGIP1
Polyclonal
PBS with 0.09% sodium azide
Q9BQI5, Q8VD37
SH3-containing GRB2-like protein 3-interacting protein 1, endophilin-3-interacting protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100752742, 73094, 84251
Western Blot
Unconjugated
SGIP1
Liquid
SGIP1
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human SGIP1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit