Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGK3, Polyclonal, Invitrogen™

Catalog No. PIPA186938
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA186938 50 μg
1 options

Catalog No. PIPA186938

Supplier: Thermo Scientific PA186938

Rabbit Polyclonal Antibody

This gene is a member of the Ser/Thr protein kinase family and encodes a phosphoprotein with a PX (phox homology) domain. The protein phosphorylates several target proteins and has a role in neutral amino acid transport and activation of potassium and chloride channels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Predicted to react with mouse based on sequence homology. Does not react with rat.Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. A suggested positive control for immunohistochemical applications is kidney.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGK3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Unconjugated
Gene SGK3
Gene Accession No. Q96BR1
Gene Alias CISK, DKFZp781N0293, SGK2, SGKL
Gene Symbols SGK3
Host Species Rabbit
Immunogen Synthetic peptide: C-terminal domain of human SGK3, conjugated to KLH
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23678
Target Species Human
Content And Storage 4°C or -20°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.