Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC5A11 Rabbit anti-Human, Monkey, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB11040436

Catalog No. NB11040436

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

SLC5A11 Polyclonal antibody specifically detects SLC5A11 in Human, Monkey samples. It is validated for Immunohistochemistry (Paraffin).
Specifications

Specifications

SLC5A11
1.0 mg/mL
Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml
Affinity Purified
SLC5A11
Synthetic 17 amino acid peptide from extracellular domain of human SLC5A11.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This is specific to the human Solute Carrier Family 5 (sodium/glucose cotransporter), Member 11 protein.
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS with 0.1% Sodium Azide
KST1RKST1, Na(+)/myo-inositol cotransporter 2, na+/myo-inositol cotransporter 2, putative sodium-coupled cotransporter RKST1, SGLT6, SLGTX, SMIT2homolog of rabbit KST1, Sodium/glucose cotransporter KST1, sodium/myo-inositol cotransporter 2, Sodium-dependent glucose cotransporter, solute carrier family 5 (sodium/glucose cotransporter), member 11, Solute carrier family 5 member 11
Rabbit
IgG
0.05 mg
Membrane Trafficking and Chaperones
Primary
115584
Human, Monkey
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only