Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC5A10, Polyclonal, Invitrogen™

Catalog No. PIPA186725
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA186725 50 μg
1 options

Catalog No. PIPA186725

Supplier: Thermo Scientific PA186725

Rabbit Polyclonal Antibody

SGLT5 is a 62kDa sodium/glucose cotransporter whose expression is restricted to kidney, and is thought to have a role in renal function. Multiple splicing transcripts exist.

Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. A suggested positive control for immunohistochemical applications is skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Unconjugated
Gene SLC5A10
Gene Accession No. A0PJK1
Gene Alias FLJ25217, SGLT5
Gene Symbols SLC5A10
Host Species Rabbit
Immunogen Synthetic peptide: Cytoplasmic domain of human SLC5A10, conjugated to KLH
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125206
Target Species Human
Content And Storage 4°C or -20°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.