Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGMS2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526802

Catalog No. PIPA526802


Add to cart

Description

Description

This antibody is predicted to react with mouse, non-human primate and rat based on sequence homology.

Sphingomyelin, a major component of cell and Golgi membranes, is made by the transfer of phosphocholine from phosphatidylcholine onto ceramide, with diacylglycerol as a side product. The protein encoded by this gene is an enzyme that catalyzes this reaction primarily at the cell membrane. The synthesis is reversible, and this enzyme can catalyze the reaction in either direction. The encoded protein is required for cell growth. Three transcript variants encoding the same protein have been found for this gene.
Specifications

Specifications

SGMS2
Polyclonal
PBS with 0.09% sodium azide
Q8NHU3
SMS2, SM synthase, phosphatidylcholine:ceramide cholinephosphotransferase 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
166929
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
SGMS2
Liquid
SGMS2
KLH conjugated synthetic peptide between 338-365 amino acids from the C-terminal region of human SGMS2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit