Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGT1 Mouse anti-Human, Unlabeled, Clone: 29, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612104

Catalog No. BDB612104


Add to cart

Description

Description

Maintenance of cellular function requires timely and selective degradation of key regulatory proteins. For example, progression of the mammalian cell cycle is regulated by the degradation of key cell cycle proteins via the ubiquitin pathway. Ubiquitin, a soluble protein of 76 amino acids, is enzymatically attached to an ε-NH2-Lys in a target protein. Ubiquitin-conjugated proteins are recognized and degraded by the 26S proteasome. Targeting of specific cellular proteins for degradation via the ubiquitylation pathway may require the formation of protein complexes. One group of ubiquitin enzyme containing complexes are the multisubunit cullin-containing RING E3 ubiquitin ligases, that include SCF and VCB-Cul-2 complexes. These complexes contain a RING domain protein, a cullin, multiple adaptor proteins, and ubiquitin enzymes. The SCF complex includes Skp1, SGT1, Cul-1, F-box protein, and CDC34/Ubc3. This complex preferentially ubiquitylates phosphoproteins, such as β-catenin, IκBα, and G1 cyclins. The mammalian SGT1 is an adaptor protein that can interact with Skp1 and can rescue yeast SGT1 null mutations. Thus, SGT1 may be an important component of the SCF ubiquination ligase complex.

Immunofluorescence, Western Blotting

Specifications

Specifications

SGT1
29
Unconjugated
Affinity Purified
Human SGT1 aa. 191-306
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit