Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGTA, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127943

Catalog No. PIPA127943

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cochaperone that binds directly to HSC70 and HSP70 and regulates their ATPase activity. This protein is a protein which is capable of interacting with the major nonstructural protein of parvovirus H-1 and 70-kDa heat shock cognate protein; however. its fu

PA1-27943 detects SGTA from human samples.PA1-27943 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Small glutamine-rich tetratricopeptide repeat-containing protein A.
Specifications

Specifications

SGTA
Polyclonal
PBS with no preservative
O43765
SGT, alphaSGT, hSGT
Chicken
IgY
50μg
Store at -20°C
Primary
6449
Western Blot
Unconjugated
SGTA
Purified
SGTA
Synthetic peptide corresponding to human Small glutamine-rich tetratricopeptide repeat-containing protein A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit